Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607659 MIM: 609237 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EAF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EAF2 - ELL associated factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320041.1 | Intron | NP_001306970.1 | ||||
NM_018456.5 | Intron | NP_060926.2 | ||||
XM_005247618.3 | Intron | XP_005247675.1 | ||||
XM_017006861.1 | Intron | XP_016862350.1 | ||||
XM_017006862.1 | Intron | XP_016862351.1 | ||||
XM_017006863.1 | Intron | XP_016862352.1 |
IQCB1 - IQ motif containing B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001023570.3 | Intron | NP_001018864.2 | ||||
NM_001023571.3 | Intron | NP_001018865.2 | ||||
NM_001319107.1 | Intron | NP_001306036.1 | ||||
XM_005247911.3 | Intron | XP_005247968.1 | ||||
XM_005247912.2 | Intron | XP_005247969.1 | ||||
XM_011513335.2 | Intron | XP_011511637.1 | ||||
XM_017007537.1 | Intron | XP_016863026.1 | ||||
XM_017007538.1 | Intron | XP_016863027.1 | ||||
XM_017007539.1 | Intron | XP_016863028.1 |