Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGTGATGGGTGGGGAAGTATCC[C/T]ATTTCTCTTAGAAAGTCTTCTGTAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164011 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105377621 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC105377621 - uncharacterized LOC105377621 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NFKB1 - nuclear factor kappa B subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165412.1 | Intron | NP_001158884.1 | ||||
NM_001319226.1 | Intron | NP_001306155.1 | ||||
NM_003998.3 | Intron | NP_003989.2 | ||||
XM_011532006.2 | Intron | XP_011530308.1 | ||||
XM_011532008.2 | Intron | XP_011530310.1 |
Set Membership: |
HapMap |