Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 607493 | |||||||||||||||||||||||
Literature Links: |
ING3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese)
|
|||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ING3 - inhibitor of growth family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019071.2 | 150 | Missense Mutation | AAC,GAC | N,D 6 | NP_061944.2 | |
NM_198267.1 | 150 | Missense Mutation | AAC,GAC | N,D 6 | NP_938008.1 | |
XM_017012369.1 | 150 | Missense Mutation | AAC,GAC | N,D 6 | XP_016867858.1 |
Set Membership: |
HapMap |