Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616292 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SAXO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SAXO1 - stabilizer of axonemal microtubules 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287049.1 | Intron | NP_001273978.1 | ||||
NM_001287050.1 | Intron | NP_001273979.1 | ||||
NM_153707.3 | Intron | NP_714918.2 | ||||
XM_011517744.1 | Intron | XP_011516046.1 | ||||
XM_011517745.1 | Intron | XP_011516047.1 | ||||
XM_011517746.2 | Intron | XP_011516048.1 | ||||
XM_011517747.2 | Intron | XP_011516049.1 | ||||
XM_017014314.1 | Intron | XP_016869803.1 | ||||
XM_017014315.1 | Intron | XP_016869804.1 |
Set Membership: |
HapMap |