Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACGGGTCCTCTCCGCGAGAAGTC[A/G]ATAATTGGCCTTTTCTTTTTTTCGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605372 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HNRNPA3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HNRNPA3 - heterogeneous nuclear ribonucleoprotein A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005246380.1 | 1371 | Intron | XP_005246437.1 | |||
XM_005246381.4 | 1371 | Intron | XP_005246438.1 | |||
XM_006712365.2 | 1371 | Intron | XP_006712428.1 | |||
XM_017003587.1 | 1371 | Intron | XP_016859076.1 | |||
XM_017003588.1 | 1371 | Intron | XP_016859077.1 |
LOC105373759 - uncharacterized LOC105373759 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017005483.1 | 1371 | Nonsense Mutation | CGA,TGA | R,* 11 | XP_016860972.1 |
MIR4444-1 - microRNA 4444-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |