Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116947 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC25A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CDC25A - cell division cycle 25A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001789.2 | 2771 | UTR 3 | NP_001780.2 | |||
NM_201567.1 | 2771 | UTR 3 | NP_963861.1 | |||
XM_006713434.2 | 2771 | UTR 3 | XP_006713497.1 | |||
XM_006713435.2 | 2771 | UTR 3 | XP_006713498.1 | |||
XM_011534316.1 | 2771 | UTR 3 | XP_011532618.1 |
Set Membership: |
HapMap |