Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 176871 | ||||||||||||||||||||
Literature Links: |
EIF2AK2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EIF2AK2 - eukaryotic translation initiation factor 2 alpha kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135651.2 | Intron | NP_001129123.1 | ||||
NM_001135652.2 | Intron | NP_001129124.1 | ||||
NM_002759.3 | Intron | NP_002750.1 | ||||
XM_011532987.2 | Intron | XP_011531289.1 | ||||
XM_017004503.1 | Intron | XP_016859992.1 |
GPATCH11 - G-patch domain containing 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |