Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604153 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC00471 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LINC00471 - long intergenic non-protein coding RNA 471 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NMUR1 - neuromedin U receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006056.4 | Intron | NP_006047.3 | ||||
XM_006712195.3 | Intron | XP_006712258.1 | ||||
XM_006712196.3 | Intron | XP_006712259.1 | ||||
XM_011510487.2 | Intron | XP_011508789.1 | ||||
XM_011510488.2 | Intron | XP_011508790.1 | ||||
XM_011510489.2 | Intron | XP_011508791.1 |
Set Membership: |
HapMap |