Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTATTATTTTTAACAATCAGAGTT[A/G]TATAACACAGGAACAGGTTGCTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
25 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDTC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
WDTC1 - WD and tetratricopeptide repeats 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276252.1 | Intron | NP_001263181.1 | ||||
NM_015023.4 | Intron | NP_055838.2 | ||||
XM_011541057.1 | Intron | XP_011539359.1 | ||||
XM_011541058.2 | Intron | XP_011539360.1 | ||||
XM_017000730.1 | Intron | XP_016856219.1 |
Set Membership: |
HapMap |