Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTTACAGTTAGGCAGAAAAATTG[A/G]ACTTATCCCAAAATGAAAGCTCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
33 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609017 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERMAP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ERMAP - erythroblast membrane associated protein (Scianna blood group) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC339539 - uncharacterized LOC339539 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF691 - zinc finger protein 691 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242739.1 | 3918 | Intron | NP_001229668.1 | |||
NM_015911.3 | 3918 | Intron | NP_056995.1 | |||
XM_006710671.3 | 3918 | Intron | XP_006710734.1 | |||
XM_017001400.1 | 3918 | Intron | XP_016856889.1 | |||
XM_017001401.1 | 3918 | Intron | XP_016856890.1 | |||
XM_017001402.1 | 3918 | UTR 5 | XP_016856891.1 | |||
XM_017001403.1 | 3918 | UTR 5 | XP_016856892.1 |
Set Membership: |
HapMap |