Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGTAAGAAAATAAAAGACAGATG[A/C]TGTAGCAAAAGGGCATTTCTCTCCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600382 MIM: 603617 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DENND4A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DENND4A - DENN domain containing 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144823.2 | 8617 | UTR 3 | NP_001138295.1 | |||
NM_001320835.1 | 8617 | UTR 3 | NP_001307764.1 | |||
NM_005848.4 | 8617 | UTR 3 | NP_005839.3 | |||
XM_005254120.4 | 8617 | Intron | XP_005254177.1 | |||
XM_005254121.3 | 8617 | Intron | XP_005254178.1 | |||
XM_005254122.4 | 8617 | Intron | XP_005254179.1 | |||
XM_006720371.3 | 8617 | Intron | XP_006720434.1 | |||
XM_011521156.2 | 8617 | Intron | XP_011519458.1 | |||
XM_011521157.2 | 8617 | Intron | XP_011519459.1 | |||
XM_017021862.1 | 8617 | Intron | XP_016877351.1 | |||
XM_017021863.1 | 8617 | Intron | XP_016877352.1 | |||
XM_017021864.1 | 8617 | Intron | XP_016877353.1 | |||
XM_017021865.1 | 8617 | Intron | XP_016877354.1 |
SLC24A1 - solute carrier family 24 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001254740.1 | 8617 | Intron | NP_001241669.1 | |||
NM_001301031.1 | 8617 | Intron | NP_001287960.1 | |||
NM_001301032.1 | 8617 | Intron | NP_001287961.1 | |||
NM_001301033.1 | 8617 | Intron | NP_001287962.1 | |||
NM_004727.2 | 8617 | Intron | NP_004718.1 | |||
XM_005254778.3 | 8617 | Intron | XP_005254835.1 | |||
XM_005254781.3 | 8617 | Intron | XP_005254838.1 | |||
XM_006720768.3 | 8617 | Intron | XP_006720831.1 | |||
XM_011522219.1 | 8617 | Intron | XP_011520521.1 | |||
XM_011522220.2 | 8617 | Intron | XP_011520522.1 | |||
XM_011522221.2 | 8617 | Intron | XP_011520523.1 | |||
XM_011522222.2 | 8617 | Intron | XP_011520524.1 | |||
XM_011522226.2 | 8617 | Intron | XP_011520528.1 | |||
XM_017022724.1 | 8617 | Intron | XP_016878213.1 | |||
XM_017022725.1 | 8617 | Intron | XP_016878214.1 |
Set Membership: |
HapMap |