Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGACCATGCAGAGGTCGCCTCTG[G/T]AAAAGGCCAGCGTTGTCTCCAAACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602421 | ||||||||||||||||||||
Literature Links: |
CFTR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CFTR - cystic fibrosis transmembrane conductance regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000492.3 | 151 | Missense Mutation | GAA,TAA | E,* 7 | NP_000483.3 | |
XM_011515751.2 | 151 | Intron | XP_011514053.1 | |||
XM_011515753.1 | 151 | Intron | XP_011514055.1 | |||
XM_011515754.2 | 151 | Intron | XP_011514056.1 | |||
XM_017011699.1 | 151 | Intron | XP_016867188.1 |