Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTATATATTTTTGTAGACAAAACA[C/T]AGGAGCTTCAGATGCCTCATAGATC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600433 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CBLN2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CBLN2 - cerebellin 2 precursor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182511.3 | 4411 | Intron | NP_872317.1 | |||
XM_006722394.3 | 4411 | UTR 3 | XP_006722457.1 | |||
XM_011525824.2 | 4411 | UTR 3 | XP_011524126.1 | |||
XM_017025559.1 | 4411 | UTR 3 | XP_016881048.1 | |||
XM_017025560.1 | 4411 | UTR 3 | XP_016881049.1 |