Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGTGGCAGACATATTTCCTCAAA[A/T]CCCATCTGATACAGGTTTAGATCCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102620 MIM: 134637 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTA2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ACTA2 - actin, alpha 2, smooth muscle, aorta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001141945.2 | 3084 | Intron | NP_001135417.1 | |||
NM_001320855.1 | 3084 | Intron | NP_001307784.1 | |||
NM_001613.2 | 3084 | Intron | NP_001604.1 |
FAS - Fas cell surface death receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000043.5 | 3084 | Intron | NP_000034.1 | |||
NM_001320619.1 | 3084 | Intron | NP_001307548.1 | |||
NM_152871.3 | 3084 | Intron | NP_690610.1 | |||
NM_152872.3 | 3084 | Intron | NP_690611.1 | |||
XM_006717819.3 | 3084 | UTR 5 | XP_006717882.1 | |||
XM_011539764.2 | 3084 | Intron | XP_011538066.1 | |||
XM_011539765.2 | 3084 | Intron | XP_011538067.1 | |||
XM_011539766.2 | 3084 | Intron | XP_011538068.1 | |||
XM_011539767.2 | 3084 | Intron | XP_011538069.1 |
Set Membership: |
HapMap |