Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCTGAGTCTCTATTTTCTTAGA[C/T]GTAAAATTGTGGGCAATCGTATGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601762 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASP10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CASP10 - caspase 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206524.1 | Intron | NP_001193453.1 | ||||
NM_001206542.1 | Intron | NP_001193471.1 | ||||
NM_001230.4 | Intron | NP_001221.2 | ||||
NM_001306083.1 | Intron | NP_001293012.1 | ||||
NM_032974.4 | Intron | NP_116756.2 | ||||
NM_032976.3 | Intron | NP_116758.1 | ||||
NM_032977.3 | Intron | NP_116759.2 | ||||
XM_005246907.2 | Intron | XP_005246964.1 | ||||
XM_006712796.3 | Intron | XP_006712859.1 | ||||
XM_011511990.2 | Intron | XP_011510292.1 |
Set Membership: |
HapMap |