Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGAGGGAAACACCTTGACATTT[G/T]ACCTTATTTCTTCTGTCTGGGTAGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611286 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RTCA PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RTCA - RNA 3'-terminal phosphate cyclase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130841.1 | Intron | NP_001124313.1 | ||||
NM_003729.3 | Intron | NP_003720.1 | ||||
XM_005271298.4 | Intron | XP_005271355.1 | ||||
XM_005271299.4 | Intron | XP_005271356.1 | ||||
XM_017002669.1 | Intron | XP_016858158.1 |
RTCA-AS1 - RTCA antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |