Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTAAACAAAATCTATCATTGCTAT[C/T]GAACAAAACAGTTGAATCAGCATGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616725 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX60L PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DDX60L - DEAD-box helicase 60-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012967.2 | 4572 | UTR 3 | NP_001012985.2 | |||
NM_001291510.1 | 4572 | Intron | NP_001278439.1 | |||
XM_005263341.4 | 4572 | UTR 3 | XP_005263398.2 | |||
XM_005263342.2 | 4572 | UTR 3 | XP_005263399.2 | |||
XM_005263343.2 | 4572 | UTR 3 | XP_005263400.2 | |||
XM_005263345.4 | 4572 | UTR 3 | XP_005263402.2 | |||
XM_006714408.2 | 4572 | UTR 3 | XP_006714471.1 | |||
XM_011532402.2 | 4572 | UTR 3 | XP_011530704.1 | |||
XM_011532403.2 | 4572 | UTR 3 | XP_011530705.1 | |||
XM_011532404.1 | 4572 | UTR 3 | XP_011530706.1 | |||
XM_011532405.1 | 4572 | UTR 3 | XP_011530707.1 | |||
XM_011532406.1 | 4572 | UTR 3 | XP_011530708.1 | |||
XM_011532409.2 | 4572 | UTR 3 | XP_011530711.1 | |||
XM_017008829.1 | 4572 | UTR 3 | XP_016864318.1 | |||
XM_017008830.1 | 4572 | UTR 3 | XP_016864319.1 | |||
XM_017008831.1 | 4572 | UTR 3 | XP_016864320.1 |