Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAACAGGCAATGTGGACTCTGTCG[C/T]GGGCATTCCCGTTCCCCTCTCTTTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
37 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608131 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUAK2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NUAK2 - NUAK family kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030952.2 | 1555 | UTR 3 | NP_112214.2 | |||
XM_005245515.4 | 1555 | UTR 3 | XP_005245572.1 |
Set Membership: |
HapMap |