Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGATTCTAGTATCTTCTTCCAAG[C/T]CACTGGTGTATAAACCTTGACAGGG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
GOT1L1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese)
|
|||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
GOT1L1 - glutamic-oxaloacetic transaminase 1-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152413.2 | 1048 | Missense Mutation | GAC,GGC | D,G 223 | NP_689626.2 | |
XM_005273399.3 | 1048 | Missense Mutation | GAC,GGC | D,G 222 | XP_005273456.1 | |
XM_006716285.3 | 1048 | Missense Mutation | GAC,GGC | D,G 192 | XP_006716348.1 | |
XM_011544391.2 | 1048 | Missense Mutation | GAC,GGC | D,G 41 | XP_011542693.1 |
Set Membership: |
HapMap |