Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGATTATATGAATTATCAGTCTC[A/G]ATAATGATTTTCCCTCCTACCTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
25 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120650 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CR2 - complement component 3d receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006658.2 | Intron | NP_001006659.1 | ||||
NM_001877.4 | Intron | NP_001868.2 | ||||
XM_011509206.2 | Intron | XP_011507508.1 |