Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCCCGCGGGGGGCGCCCGGGGCT[C/G]GGGGGGCGGCCCGCCGCCGTGGCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609171 MIM: 616575 | ||||||||||||||||||||
Literature Links: |
CDC42EP5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDC42EP5 - CDC42 effector protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145057.3 | 543 | Missense Mutation | CAG,GAG | Q,E 59 | NP_659494.2 |
LENG8 - leukocyte receptor cluster member 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LENG9 - leukocyte receptor cluster member 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |