Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGCTGTGGGCCAAATCCCCTGGG[A/G]TACTGGCAGGGCAGCCTTTCTTCGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606248 | ||||||||||||||||||||
Literature Links: |
QPRT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
QPRT - quinolinate phosphoribosyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318249.1 | 347 | Intron | NP_001305178.1 | |||
NM_001318250.1 | 347 | Intron | NP_001305179.1 | |||
NM_014298.4 | 347 | Missense Mutation | ATA,GTA | I,V 53 | NP_055113.2 | |
XM_005255223.3 | 347 | Intron | XP_005255280.2 | |||
XM_017023101.1 | 347 | Intron | XP_016878590.1 |