Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTCGTTGCCCCTGGGGTCATCGC[A/T]CCCCGCCGAACATCAAATGATTCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606280 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCAF16 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DCAF16 - DDB1 and CUL4 associated factor 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017741.3 | Intron | NP_060211.3 | ||||
XM_005248169.3 | Intron | XP_005248226.1 | ||||
XM_005248171.3 | Intron | XP_005248228.1 | ||||
XM_006713966.3 | Intron | XP_006714029.1 | ||||
XM_017008327.1 | Intron | XP_016863816.1 | ||||
XM_017008328.1 | Intron | XP_016863817.1 |
NCAPG - non-SMC condensin I complex subunit G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022346.4 | Intron | NP_071741.2 | ||||
XM_011513876.2 | Intron | XP_011512178.1 | ||||
XM_017008543.1 | Intron | XP_016864032.1 | ||||
XM_017008544.1 | Intron | XP_016864033.1 |
Set Membership: |
HapMap |