Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACATATATGCTATCTAGAGCCCAT[C/T]TACTCTATGTCAGCTTTGGCTTCTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604397 MIM: 605978 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNA14 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GNA14 - G protein subunit alpha 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004297.3 | 2241 | UTR 3 | NP_004288.1 | |||
XM_011519224.1 | 2241 | UTR 3 | XP_011517526.1 | |||
XM_017015341.1 | 2241 | UTR 3 | XP_016870830.1 |
VPS13A - vacuolar protein sorting 13 homolog A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |