Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAAGTCGCTGCCTAGGATGGCCA[AT/GC]TGGTCCCTGGCGATGAGCAGTGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604996 MIM: 608303 | ||||||||||||||||||||
Literature Links: |
FXYD3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FXYD3 - FXYD domain containing ion transport regulator 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LGI4 - leucine rich repeat LGI family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139284.2 | 1390 | Missense Mutation | CAATCT,CAGCTG | Q,L S465Q | NP_644813.1 | |
XM_011526595.1 | 1390 | Missense Mutation | CAATCT,CAGCTG | Q,L S293Q | XP_011524897.1 | |
XM_017026428.1 | 1390 | Missense Mutation | CAATCT,CAGCTG | Q,L S293Q | XP_016881917.1 | |
XM_017026429.1 | 1390 | Missense Mutation | CAATCT,CAGCTG | Q,L S293Q | XP_016881918.1 | |
XM_017026430.1 | 1390 | Missense Mutation | CAATCT,CAGCTG | Q,L S293Q | XP_016881919.1 |
MIR6887 - microRNA 6887 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |