Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGCCCCCACAACCCAAGTATCCA[C/T]AGAGAGTGTGCAGGAGCGGGGACCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610296 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUDCD3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NUDCD3 - NudC domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015332.3 | 2671 | UTR 3 | NP_056147.2 | |||
XM_011515247.2 | 2671 | Intron | XP_011513549.1 | |||
XM_017011908.1 | 2671 | Intron | XP_016867397.1 | |||
XM_017011909.1 | 2671 | UTR 3 | XP_016867398.1 | |||
XM_017011910.1 | 2671 | UTR 3 | XP_016867399.1 |
Set Membership: |
HapMap |