Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGATGAGCGCTTACCAACTCTTG[C/T]AATATTAGGTGATAGGACTTTGTGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
24 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613173 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NEGR1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NEGR1 - neuronal growth regulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173808.2 | 2208 | UTR 3 | NP_776169.2 | |||
XM_011541200.2 | 2208 | Intron | XP_011539502.1 | |||
XM_011541201.2 | 2208 | Intron | XP_011539503.1 | |||
XM_017000961.1 | 2208 | Intron | XP_016856450.1 |
Set Membership: |
HapMap |