Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCCTACGGCCCCTGAATATTTTG[A/G]CATCTTCAACCTACCGCAACTGTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611841 MIM: 609139 | ||||||||||||||||||||
Literature Links: |
MRPL35 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MRPL35 - mitochondrial ribosomal protein L35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016622.3 | 128 | Missense Mutation | ACA,GCA | T,A 24 | NP_057706.2 | |
NM_145644.2 | 128 | Missense Mutation | ACA,GCA | T,A 24 | NP_663619.1 | |
XM_005264361.2 | 128 | Intron | XP_005264418.1 |
REEP1 - receptor accessory protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |