Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGTGAACAGGGCCAAACGGAAAG[C/T]AGGGGTGGATCCTCTGGTACCGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606828 | ||||||||||||||||||||
Literature Links: |
NAGK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NAGK - N-acetylglucosamine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017567.4 | 550 | Missense Mutation | GCA,GTA | A,V 106 | NP_060037.3 | |
XM_005264412.4 | 550 | Missense Mutation | GCA,GTA | A,V 9 | XP_005264469.1 | |
XM_005264413.1 | 550 | Missense Mutation | GCA,GTA | A,V 9 | XP_005264470.1 | |
XM_005264415.2 | 550 | Missense Mutation | GCA,GTA | A,V 9 | XP_005264472.1 | |
XM_011532957.1 | 550 | Missense Mutation | GCA,GTA | A,V 9 | XP_011531259.1 | |
XM_017004440.1 | 550 | Missense Mutation | GCA,GTA | A,V 106 | XP_016859929.1 |