Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAATTTGCTTCCTCACAGATGACA[C/T]TGCTGGTTTATAAGGGGTACTTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601478 MIM: 616496 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYO1A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MYO1A - myosin IA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEMP1 - nuclear envelope integral membrane protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130963.1 | 2735 | UTR 3 | NP_001124435.1 | |||
NM_015257.2 | 2735 | UTR 3 | NP_056072.2 | |||
XM_011538056.2 | 2735 | UTR 3 | XP_011536358.1 | |||
XM_011538057.2 | 2735 | UTR 3 | XP_011536359.1 | |||
XM_011538058.2 | 2735 | UTR 3 | XP_011536360.1 | |||
XM_011538059.1 | 2735 | UTR 3 | XP_011536361.1 | |||
XM_011538060.2 | 2735 | Intron | XP_011536362.1 | |||
XM_017019077.1 | 2735 | UTR 3 | XP_016874566.1 | |||
XM_017019078.1 | 2735 | UTR 3 | XP_016874567.1 | |||
XM_017019079.1 | 2735 | UTR 3 | XP_016874568.1 |