Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCAAGGCTGAAGATGTTTTGGAT[A/G]TTTGGATGACACCGACGTACCCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
32 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614612 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KANK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KANK4 - KN motif and ankyrin repeat domains 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320269.1 | 1882 | UTR 3 | NP_001307198.1 | |||
NM_181712.4 | 1882 | UTR 3 | NP_859063.3 | |||
XM_011540843.2 | 1882 | UTR 3 | XP_011539145.1 | |||
XM_011540845.2 | 1882 | UTR 3 | XP_011539147.1 | |||
XM_017000483.1 | 1882 | UTR 3 | XP_016855972.1 | |||
XM_017000484.1 | 1882 | UTR 3 | XP_016855973.1 | |||
XM_017000485.1 | 1882 | UTR 3 | XP_016855974.1 |
Set Membership: |
HapMap |