Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTTAAAGGTGGTGCCTTTTCAGG[C/T]AATGTCAACTGGCCAGTTCTGTTGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604417 MIM: 601918 MIM: 611373 MIM: 612080 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AFF4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AFF4 - AF4/FMR2 family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014423.3 | 8860 | UTR 3 | NP_055238.1 | |||
XM_005271963.4 | 8860 | UTR 3 | XP_005272020.1 | |||
XM_005271964.4 | 8860 | UTR 3 | XP_005272021.1 | |||
XM_006714587.3 | 8860 | UTR 3 | XP_006714650.1 |
GDF9 - growth differentiation factor 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LEAP2 - liver enriched antimicrobial peptide 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UQCRQ - ubiquinol-cytochrome c reductase complex III subunit VII | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |