Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGGGAGGACACCCAGCACTGGCA[C/T]TGGGAGAACTCCAGATTCCCTACAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609987 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STRA8 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
STRA8 - stimulated by retinoic acid 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182489.1 | Intron | NP_872295.1 | ||||
XM_011516137.2 | Intron | XP_011514439.1 | ||||
XM_011516138.2 | Intron | XP_011514440.1 | ||||
XM_011516139.2 | Intron | XP_011514441.1 | ||||
XM_017012150.1 | Intron | XP_016867639.1 |