Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATCCGCACAATAACTCTGTGAAG[C/G]AGATAGTATCATTCCATTTTATAGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612803 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928865 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC101928865 - uncharacterized LOC101928865 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NARS2 - asparaginyl-tRNA synthetase 2, mitochondrial (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243251.1 | Intron | NP_001230180.1 | ||||
NM_024678.5 | Intron | NP_078954.4 | ||||
XM_011545253.1 | Intron | XP_011543555.1 | ||||
XM_017018302.1 | Intron | XP_016873791.1 | ||||
XM_017018303.1 | Intron | XP_016873792.1 | ||||
XM_017018304.1 | Intron | XP_016873793.1 |
Set Membership: |
HapMap |