Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGATGTAAAATAAATTAAGGAAGA[A/G]ATATTTAGCACAATATGGAATGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601810 MIM: 610328 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNA2 - DNA replication helicase/nuclease 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080449.2 | Intron | NP_001073918.2 | ||||
XM_006717680.3 | Intron | XP_006717743.1 | ||||
XM_011539417.1 | Intron | XP_011537719.1 | ||||
XM_017015799.1 | Intron | XP_016871288.1 |
RUFY2 - RUN and FYVE domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |