Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602926 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP157 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CFAP157 - cilia and flagella associated protein 157 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3911 - microRNA 3911 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTRH1 - peptidyl-tRNA hydrolase 1 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002913.1 | Intron | NP_001002913.1 | ||||
XM_006716955.3 | Intron | XP_006717018.1 | ||||
XM_017014276.1 | Intron | XP_016869765.1 | ||||
XM_017014277.1 | Intron | XP_016869766.1 | ||||
XM_017014278.1 | Intron | XP_016869767.1 |
STXBP1 - syntaxin binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |