Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCCGGCGCCTTTCCGGGCCATTC[G/A]GCGGGGCACGACAAACGGCAGGTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 160775 | ||||||||||||||||||||
Literature Links: |
MIR6819 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR6819 - microRNA 6819 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYH9 - myosin, heavy chain 9, non-muscle | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002473.5 | 5832 | Nonsense Mutation | CGA,TGA | R,* 1933 | NP_002464.1 | |
XM_011530197.2 | 5832 | Nonsense Mutation | CGA,TGA | R,* 1933 | XP_011528499.1 | |
XM_017028803.1 | 5832 | Nonsense Mutation | CGA,TGA | R,* 1933 | XP_016884292.1 | |
XM_017028804.1 | 5832 | Nonsense Mutation | CGA,TGA | R,* 1933 | XP_016884293.1 | |
XM_017028805.1 | 5832 | Nonsense Mutation | CGA,TGA | R,* 1933 | XP_016884294.1 | |
XM_017028806.1 | 5832 | Nonsense Mutation | CGA,TGA | R,* 1933 | XP_016884295.1 |