Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGAGAGAGAAAACATTATGTAC[A/G]TGTGTTTGGCAAGCATATTCTCTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300976 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHMP1B2P PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CHMP1B2P - charged multivesicular body protein 1B2, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM46D - family with sequence similarity 46 member D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170574.1 | Intron | NP_001164045.1 | ||||
NM_152630.4 | Intron | NP_689843.1 |