Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGTGGGTTTTTAGCGTTGCTTAT[A/G]TATTCTTTAACTCCAGTTATTTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603216 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAPG PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NAPG - NSF attachment protein gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003826.2 | Intron | NP_003817.1 | ||||
XM_011525754.2 | Intron | XP_011524056.1 | ||||
XM_011525756.2 | Intron | XP_011524058.1 | ||||
XM_017026063.1 | Intron | XP_016881552.1 |
Set Membership: |
HapMap |