Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAATGTTGAGTAATGAATTTAAA[C/G]TGTTTTTAACATTGACCCTCCATTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603431 MIM: 615070 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF4H PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
EIF4H - eukaryotic translation initiation factor 4H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022170.1 | Intron | NP_071496.1 | ||||
NM_031992.1 | Intron | NP_114381.1 | ||||
XM_011516556.1 | Intron | XP_011514858.1 | ||||
XM_011516557.1 | Intron | XP_011514859.1 |
MIR590 - microRNA 590 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |