Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTAAGGGAGGCTACAGGGATATA[C/T]TGACCCCAACCACACATCTACAGAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NKPD1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NKPD1 - NTPase, KAP family P-loop domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198478.3 | 5921 | UTR 3 | NP_940880.3 | |||
XM_011526799.2 | 5921 | UTR 3 | XP_011525101.1 | |||
XM_011526802.2 | 5921 | UTR 3 | XP_011525104.1 | |||
XM_011526804.2 | 5921 | UTR 3 | XP_011525106.1 | |||
XM_011526805.2 | 5921 | UTR 3 | XP_011525107.1 |
PPP1R37 - protein phosphatase 1 regulatory subunit 37 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |