Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGGTTGCAGGTGGGGGCAAAGC[A/G]TGCAGACCGCTGTCAGCACAAAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610190 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHST8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CHST8 - carbohydrate sulfotransferase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127895.1 | Intron | NP_001121367.1 | ||||
NM_001127896.1 | Intron | NP_001121368.1 | ||||
NM_022467.3 | Intron | NP_071912.2 | ||||
XM_011527222.1 | Intron | XP_011525524.1 | ||||
XM_011527224.1 | Intron | XP_011525526.1 | ||||
XM_017027143.1 | Intron | XP_016882632.1 |
Set Membership: |
HapMap |