Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCGTATACTACATGAGGAATTTC[A/G]GCTTTATGGCATTAGTCATTTCCAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607746 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FERMT2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FERMT2 - fermitin family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134999.1 | Intron | NP_001128471.1 | ||||
NM_001135000.1 | Intron | NP_001128472.1 | ||||
NM_006832.2 | Intron | NP_006823.1 | ||||
XM_005267285.2 | Intron | XP_005267342.1 | ||||
XM_006720008.2 | Intron | XP_006720071.1 | ||||
XM_006720009.2 | Intron | XP_006720072.1 | ||||
XM_006720010.2 | Intron | XP_006720073.1 |