Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGCTGTCCATGTGCAGCTGTCTAA[C/T]GTTAGGCATTACAAAGCTAAATTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600381 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KTN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KTN1 - kinectin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079521.1 | 3573 | Intron | NP_001072989.1 | |||
NM_001079522.1 | 3573 | Intron | NP_001072990.1 | |||
NM_001271014.1 | 3573 | Intron | NP_001257943.1 | |||
NM_004986.3 | 3573 | Intron | NP_004977.2 | |||
XM_006720138.2 | 3573 | Intron | XP_006720201.1 | |||
XM_006720139.2 | 3573 | Intron | XP_006720202.1 | |||
XM_006720140.3 | 3573 | Intron | XP_006720203.1 | |||
XM_006720141.2 | 3573 | Intron | XP_006720204.1 | |||
XM_011536751.2 | 3573 | Intron | XP_011535053.1 | |||
XM_011536752.1 | 3573 | Intron | XP_011535054.1 | |||
XM_011536753.1 | 3573 | Intron | XP_011535055.1 | |||
XM_017021278.1 | 3573 | UTR 5 | XP_016876767.1 | |||
XM_017021279.1 | 3573 | Intron | XP_016876768.1 | |||
XM_017021280.1 | 3573 | Intron | XP_016876769.1 | |||
XM_017021281.1 | 3573 | UTR 5 | XP_016876770.1 | |||
XM_017021282.1 | 3573 | Intron | XP_016876771.1 | |||
XM_017021283.1 | 3573 | Intron | XP_016876772.1 | |||
XM_017021284.1 | 3573 | Intron | XP_016876773.1 | |||
XM_017021285.1 | 3573 | Intron | XP_016876774.1 | |||
XM_017021286.1 | 3573 | Intron | XP_016876775.1 |
KTN1-AS1 - KTN1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |