Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCAGTCTGTGACCTCGGCCTTA[A/G]GCTAATCATTCTTTGGTTCACATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608986 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRTC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CRTC3 - CREB regulated transcription coactivator 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042574.2 | Intron | NP_001036039.1 | ||||
NM_022769.4 | Intron | NP_073606.3 | ||||
XM_005254968.2 | Intron | XP_005255025.1 | ||||
XM_011521906.2 | Intron | XP_011520208.1 |
Set Membership: |
HapMap |