Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTAGTATGATTGTGCTTACATCT[A/G]CCCCATATATGTAAGTTTTATTATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604961 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDE11A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PDE11A - phosphodiesterase 11A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077196.1 | 5854 | UTR 3 | NP_001070664.1 | |||
NM_001077197.1 | 5854 | UTR 3 | NP_001070665.1 | |||
NM_001077358.1 | 5854 | Intron | NP_001070826.1 | |||
NM_016953.3 | 5854 | UTR 3 | NP_058649.3 |
TTC30A - tetratricopeptide repeat domain 30A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |