Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTTAAAAAAGGAAACCAAGTACC[A/G]TTTGTCTCCATCAACACCAGGCACT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605292 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NT5M PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
NT5M - 5',3'-nucleotidase, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020201.3 | Intron | NP_064586.1 | ||||
XM_005256731.3 | Intron | XP_005256788.1 | ||||
XM_005256732.3 | Intron | XP_005256789.1 | ||||
XM_005256733.3 | Intron | XP_005256790.1 | ||||
XM_005256734.3 | Intron | XP_005256791.1 | ||||
XM_005256737.1 | Intron | XP_005256794.1 | ||||
XM_011523962.2 | Intron | XP_011522264.1 | ||||
XM_011523963.1 | Intron | XP_011522265.1 | ||||
XM_011523964.2 | Intron | XP_011522266.1 | ||||
XM_011523965.2 | Intron | XP_011522267.1 | ||||
XM_011523966.1 | Intron | XP_011522268.1 | ||||
XM_011523967.2 | Intron | XP_011522269.1 | ||||
XM_017024879.1 | Intron | XP_016880368.1 |