Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTAAGGGTTCATCTTTAAATAAAA[C/T]TGATACTAAATCAATTATTCCATTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602848 MIM: 603575 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRD8 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BRD8 - bromodomain containing 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NME5 - NME/NM23 family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003551.2 | Intron | NP_003542.1 | ||||
XM_005272099.1 | Intron | XP_005272156.1 | ||||
XM_006714716.3 | Intron | XP_006714779.1 | ||||
XM_017009945.1 | Intron | XP_016865434.1 |
Set Membership: |
HapMap |