Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGACTGTGGCCCCAGCCTCTGAGA[C/G]AAAGGACAGTGGAGGCCCCGGGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
26 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601749 MIM: 611476 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLMN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GLMN - glomulin, FKBP associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319683.1 | Intron | NP_001306612.1 | ||||
NM_053274.2 | Intron | NP_444504.1 | ||||
XM_005270401.3 | Intron | XP_005270458.1 | ||||
XM_006710309.2 | Intron | XP_006710372.1 | ||||
XM_011540546.2 | Intron | XP_011538848.1 | ||||
XM_017000137.1 | Intron | XP_016855626.1 | ||||
XM_017000138.1 | Intron | XP_016855627.1 | ||||
XM_017000139.1 | Intron | XP_016855628.1 | ||||
XM_017000140.1 | Intron | XP_016855629.1 | ||||
XM_017000141.1 | Intron | XP_016855630.1 | ||||
XM_017000142.1 | Intron | XP_016855631.1 | ||||
XM_017000143.1 | Intron | XP_016855632.1 | ||||
XM_017000144.1 | Intron | XP_016855633.1 |
RPAP2 - RNA polymerase II associated protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024813.2 | Intron | NP_079089.2 | ||||
XM_005271223.2 | Intron | XP_005271280.1 | ||||
XM_011542167.2 | Intron | XP_011540469.1 | ||||
XM_017002362.1 | Intron | XP_016857851.1 | ||||
XM_017002363.1 | Intron | XP_016857852.1 | ||||
XM_017002364.1 | Intron | XP_016857853.1 |
Set Membership: |
HapMap |