Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTGTCTATTGTCTGTTTCAGCA[C/T]TGAAGGGGTAAGAATATTTTTCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603666 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STX16 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
STX16 - syntaxin 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001433.2 | Intron | NP_001001433.1 | ||||
NM_001134772.2 | Intron | NP_001128244.1 | ||||
NM_001134773.2 | Intron | NP_001128245.1 | ||||
NM_001204868.1 | Intron | NP_001191797.1 | ||||
NM_003763.5 | Intron | NP_003754.2 |
STX16-NPEPL1 - STX16-NPEPL1 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |